Learn More
Cayman Chemical Edoxaban (tosylate), Purity: ≥98%, CAS: 480449-71-6, 5 mg
SDP
Catalog No. 502933259
Encompass_Preferred
An orally bioavailable and potent inhibitor of factor Xa (Kis = 0.561 and 0.903 nM for free and complexed human factor Xa, respectively); >10,000-fold selective for factor Xa over thrombin, FVIIa/sTF, FXIa, tPA, aPC, trypsin, plasmin, and chymotrypsin; prolongs prothrombin, activated partial thromboplastin, and thrombin clotting times in a concentration-dependent manner ex vivo in human plasma; reduces thrombus formation and prolongs prothrombin time in a dose-dependent manner in rat and rabbit models of venous stasis thrombosis when administered at doses ranging from 0.5-12.5 mg/kg; reduces thrombus formation in a rat model of platinum wire-induced venous thrombosis
Catalog No. 50-293-3259 Supplier Cayman Chemical Supplier No. 240305 MG
An orally bioavailable and potent inhibitor of factor Xa (Kis = 0.561 and 0.903 nM for free and complexed human factor Xa, respectively); >10,000-fold selective for factor Xa over thrombin, FVIIa/sTF, FXIa, tPA, aPC, trypsin, plasmin, and chymotrypsin; prolongs prothrombin, activated partial thromboplastin, and thrombin clotting times in a concentration-dependent manner ex vivo in human plasma; reduces thrombus formation and prolongs prothrombin time in a dose-dependent manner in rat and rabbit models of venous stasis thrombosis when administered at doses ranging from 0.5-12.5 mg/kg; reduces thrombus formation in a rat model of platinum wire-induced venous thrombosis