Learn More
Pipette Supplies, Inc. EDP2 Battery, Lithium 3.6 Volt
SDP
Catalog No. 501317651
Encompass
Same batteries as sold by Rainin. LSH-14, Saft “C” cell.3.6V, 5.8 Ah, 1.7g lithium-metal content. Formerly RA E2-BA. Please note that Pipette Supplies only ships this battery via Ground Shipping. Made in France
Catalog No. 50-131-7651 Supplier Pipette Supplies, Inc. Supplier No. PS E2BA
Same batteries as sold by Rainin. LSH-14, Saft “C” cell.3.6V, 5.8 Ah, 1.7g lithium-metal content. Formerly RA E2-BA. Please note that Pipette Supplies only ships this battery via Ground Shipping. Made in France