Learn More
TARGETMOL CHEMICALS INC EDRALBRUTINIB 5MG
SDP
Catalog No. 502579429
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Edralbrutinib (TG-1701) is a potent BTK inhibitor with anticancer activity and is used in the treatment of tumors immune system disorders and blood and lymphatic system disorders.Edralbrutinib is used in the study of membranous glomerulonephritis and optic neuromyelitis optica. purity: 99%
Catalog No. 50-257-9429 Supplier TARGETMOL CHEMICALS INC Supplier No. T632725MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Edralbrutinib (TG-1701) is a potent BTK inhibitor with anticancer activity and is used in the treatment of tumors immune system disorders and blood and lymphatic system disorders.Edralbrutinib is used in the study of membranous glomerulonephritis and optic neuromyelitis optica. purity: 99%