Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Edrophonium chloride | 116-38-1 | 99.4% | 1 ML
SDP

Catalog No. 5000408664
Encompass_Preferred

Edrophonium chloride is a short-acting acetylcholinesterase (AChE) inhibitor with a molecular formula of C10H16ClNO and a molecular weight of 201.69. It exhibits Ki values of 0.2, 0.2, and 0.4 μM, and IC50 values of 0.2, 0.05, and 0.5 μM for AChE derived from human erythrocytes, purified calf forebrain, and purified octopus brain, respectively. This compound is utilized in medical contexts for the detection of early digitalis toxicity and in studies related to myasthenia gravis.

  • Short-acting acetylcholinesterase inhibitor
  • Formula: C10H16ClNO
  • Molecular weight: 201.69
  • Appearance: Solid
  • Melting/freezing point: 162-163°C
  • Relative density: 1.0787 g/cm3
  • Identified uses include laboratory chemicals and manufacture of substances

Catalog No. 50-004-08664 Supplier Medchemexpress LLC Supplier No. HYB088210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Edrophonium chloride is a short-acting acetylcholinesterase (AChE) inhibitor with a molecular formula of C10H16ClNO and a molecular weight of 201.69. It exhibits Ki values of 0.2, 0.2, and 0.4 μM, and IC50 values of 0.2, 0.05, and 0.5 μM for AChE derived from human erythrocytes, purified calf forebrain, and purified octopus brain, respectively. This compound is utilized in medical contexts for the detection of early digitalis toxicity and in studies related to myasthenia gravis.

  • Short-acting acetylcholinesterase inhibitor
  • Formula: C10H16ClNO
  • Molecular weight: 201.69
  • Appearance: Solid
  • Melting/freezing point: 162-163°C
  • Relative density: 1.0787 g/cm3
  • Identified uses include laboratory chemicals and manufacture of substances

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.