Learn More
Cayman Chemical Edrophonium (chloride), Purity: ≥95%, CAS: 116-38-1, 100 mg
SDP
Catalog No. 502968681
Encompass_Preferred
An AChE inhibitor (Kis = 0.2 - 0.4 µM) that is known to prevent the breakdown of the neurotransmitter acetylcholine by binding specifically to its catalytic site; used as part of a battery of pharmacological tests to confirm a diagnoses of myasthenia gravis
Catalog No. 50-296-8681 Supplier Cayman Chemical Supplier No. 15928100 MG
An AChE inhibitor (Kis = 0.2 - 0.4 µM) that is known to prevent the breakdown of the neurotransmitter acetylcholine by binding specifically to its catalytic site; used as part of a battery of pharmacological tests to confirm a diagnoses of myasthenia gravis