Learn More
EDTA, 0.0010 Molar, Reagents
EDTA, 0.0010 Molar, 1 mL = 0.1 mg CaCO3, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$168.00
Chemical Identifiers
| CAS | 7732-18-5 , 6381-92-6 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-335-352
|
Reagents Holdings Llc
CE25080010F |
10 L |
Each of 1 for $168.00
|
|
|||||
Chemical Identifiers
| 7732-18-5 , 6381-92-6 | |
| 18.02 | |
| Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| Colorless | |
| 100°C | |
| Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt | |
| O | |
| 18.02 | |
| Cubitainer | |
| 4 to 6 | |
| Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.