Learn More
Hach Company EDTA Digital Titrator Cartridge, 0.1428 M
Catalog No. 501995152 Shop All Hach Company Products
Encompass
For the determination of Calcium and Total Hardness. 0.1428 M. Digits x 0.1785 = mg/L CaCO3 in 100 mL, Digits x 0.01 = Gdh.
Catalog No. 50-199-5152 Supplier Hach Company Supplier No. 1496001
For the determination of Calcium and Total Hardness. 0.1428 M. Digits x 0.1785 = mg/L CaCO3 in 100 mL, Digits x 0.01 = Gdh.