Learn More
Aqua Solutions EDTA 0.1N
SDP
Catalog No. NC9732027
Encompass_Preferred
Edta Titrant 0.1 Normal (0.05m) N.i.s.t. Traceable Prepared To Astm E200 (apha)
Catalog No. NC9732027 Supplier Aqua Solutions Supplier No. 36401L
Edta Titrant 0.1 Normal (0.05m) N.i.s.t. Traceable Prepared To Astm E200 (apha)