Learn More
Boston Bioproducts Inc EDTA (0.5 M, pH 7.4, pH Adjusted with KOH) - 250ml
Catalog No. NC1904325
Encompass
EDTA [Ethylenediaminetetraacetic acid] solution is commonly used in various molecular biology and protein biochemistry applications that require the chelation of divalent metal cations, such as calcium (Ca2+) and magnesium (Mg2+). EDTA is used extensively as an anticoagulant in blood samples as it chelates the calcium present, preventing coagulation and preserving blood cell morphology. It is also used in cell lysis buffers. This solution is adjusted with KOH.
Catalog No. NC1904325 Supplier Boston Bioproducts Inc Supplier No. BM711K250ML
EDTA [Ethylenediaminetetraacetic acid] solution is commonly used in various molecular biology and protein biochemistry applications that require the chelation of divalent metal cations, such as calcium (Ca2+) and magnesium (Mg2+). EDTA is used extensively as an anticoagulant in blood samples as it chelates the calcium present, preventing coagulation and preserving blood cell morphology. It is also used in cell lysis buffers. This solution is adjusted with KOH.