Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc EDTA (0.5 M), pH 8.0, PK 4 X 100 mL
SDP

Catalog No. 50983252
Encompass_Preferred

EDTA (Ethylenediaminetetraacetic acid), a metal chelator, is widely used in molecular biology as a chemical reagent to reduce nuclease activity (i.e. metalloenzyme). EDTA solution can be used to study cell biology, molecular biology, bioactive small molecules, biochemicals, solutions and reagents, inorganic salts, and chelators.

Applications: Typically, EDTA is added to Tris base in order to prepare Tris EDTA (TE), which can be used to reconstitute nucleic acids (i.e. DNA).

Storage Conditions: 15 °C to 30°C

Shipping Condition: ambient temperature Sterile Filtered
DNase, RNase and Protease tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-983-252 Supplier Quality Biological Inc Supplier No. 351027721
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EDTA (Ethylenediaminetetraacetic acid), a metal chelator, is widely used in molecular biology as a chemical reagent to reduce nuclease activity (i.e. metalloenzyme). EDTA solution can be used to study cell biology, molecular biology, bioactive small molecules, biochemicals, solutions and reagents, inorganic salts, and chelators.

Applications: Typically, EDTA is added to Tris base in order to prepare Tris EDTA (TE), which can be used to reconstitute nucleic acids (i.e. DNA).

Storage Conditions: 15 °C to 30°C

Shipping Condition: ambient temperature Sterile Filtered
DNase, RNase and Protease tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.