Learn More
VWR INTERNATIONAL LLC EDTA, 0.5M STERILE SOLUTION, pH 8.0, 100 mL
Catalog No. NC9942732
Encompass
UltraPure 0.5 M EDTA, pH 8.0 is an aqueous solution prepared by dissolving Na2EDTA•2H2O in distilled, deionized water and adjusting the pH to 8.0 with sodium hydroxide. This sterile-filtered solution is suitable for biochemistry or molecular biology applications requiring a chelator of divalent metal ions.
Catalog No. NC9942732 Supplier VWR INTERNATIONAL LLC Supplier No. 97062836
Add to Cart
Add to Cart
UltraPure 0.5 M EDTA, pH 8.0 is an aqueous solution prepared by dissolving Na2EDTA•2H2O in distilled, deionized water and adjusting the pH to 8.0 with sodium hydroxide. This sterile-filtered solution is suitable for biochemistry or molecular biology applications requiring a chelator of divalent metal ions.