Learn More
Alkali Scientific EDTA 0.5M Solution pH 8.0, 500 Milliliters
SDP
Catalog No. 5000660317
Encompass_Preferred
EDTA (Ethylenediaminetetraacetic Acid) 0.5M solution at pH 8.0 is a potent chelating agent used in molecular biology and biochemistry to bind metal ions that could catalyze the degradation of nucleic acids or proteins. This 500mL solution is typically used in large-scale applications like DNA extraction, RNA stabilization, and protein purification, where controlling metal ion presence is crucial for preserving sample integrity. The 0.5M concentration and pH 8.0 provide optimal conditions for the stability of biological samples during storage and processing. EDTA is also used in laboratory buffers and reagents for routine biochemical assays, ensuring accurate, reproducible results in various experimental protocols.
Catalog No. 50-006-60317 Supplier Alkali Scientific Supplier No. EDT003CP
EDTA (Ethylenediaminetetraacetic Acid) 0.5M solution at pH 8.0 is a potent chelating agent used in molecular biology and biochemistry to bind metal ions that could catalyze the degradation of nucleic acids or proteins. This 500mL solution is typically used in large-scale applications like DNA extraction, RNA stabilization, and protein purification, where controlling metal ion presence is crucial for preserving sample integrity. The 0.5M concentration and pH 8.0 provide optimal conditions for the stability of biological samples during storage and processing. EDTA is also used in laboratory buffers and reagents for routine biochemical assays, ensuring accurate, reproducible results in various experimental protocols.