Learn More
Research Products International Corp EDTA 0.5M Solution pH 8.0, 100 Milliliters

Supplier: Research Products International Corp E14000100.0
- Soluble in: Water
- Store at: 2-8°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.