Learn More
Hach Company EDTA Digital Titrator Cartridge, 0.714 M
Supplier:  Hach Company 1495901
Encompass
EDTA Cartridge for Digital Titrator for the determination of Total Hardness, 0.714 M. Digits x 1.785 = mg/L CaCO3 in 50 mL, Digits x 0.1 = Gdh.
Catalog No. 50-199-5151