Learn More
SAI Infusion Technologies 1.3ml K3 EDTA Blood Tubes with Flip Cap
SDP
Catalog No. NC0826920
Encompass
1.3ml K3EDTA Blood Tube with attached flip cap. Ideal for collection of up to 1.3ml of blood for plasma. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/pk. Product is for laboratory animal research only.
Catalog No. NC0826920 Supplier SAI Infusion Technologies Supplier No. PMTPE1.3
1.3ml K3EDTA Blood Tube with attached flip cap. Ideal for collection of up to 1.3ml of blood for plasma. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/pk. Product is for laboratory animal research only.