Learn More
ILEX LIFE SCIENCES LLC EDTA 1P P10-026100 100 ML
SDP
Catalog No. 502733669
Encompass_Preferred
PAN-Biotech EDTA 1% in PBS, w/o: Ca and Mg (100 ml). DESCRIPTION: PAN-Biotech EDTA 1% in PBS, w/o: Ca and Mg. Ethylenediaminetetraacetic acid (EDTA), a chelating agent, is often used to remove calcium from cell washing and suspension media. EDTA can reduce clumping of cells grown in liquid suspension. It is also used for gentle detachment of certain adherent cells for passaging. EDTA is often added to trypsin solutions to enhance enzymatic activity by neutralizing calcium and magnesium ions that enhance cell-to-cell adhesion and obscure the peptide bonds on which trypsin acts. LIQUID/POWDER: Liquid. PHENOL RED: Without Phenol Red. Ca/Mg CONTENT: Without Ca and Mg. EDTA: 1.00% EDTA. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH (Germany).
Catalog No. 50-273-3669 Supplier ILEX LIFE SCIENCES LLC Supplier No. P10026100
PAN-Biotech EDTA 1% in PBS, w/o: Ca and Mg (100 ml). DESCRIPTION: PAN-Biotech EDTA 1% in PBS, w/o: Ca and Mg. Ethylenediaminetetraacetic acid (EDTA), a chelating agent, is often used to remove calcium from cell washing and suspension media. EDTA can reduce clumping of cells grown in liquid suspension. It is also used for gentle detachment of certain adherent cells for passaging. EDTA is often added to trypsin solutions to enhance enzymatic activity by neutralizing calcium and magnesium ions that enhance cell-to-cell adhesion and obscure the peptide bonds on which trypsin acts. LIQUID/POWDER: Liquid. PHENOL RED: Without Phenol Red. Ca/Mg CONTENT: Without Ca and Mg. EDTA: 1.00% EDTA. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH (Germany).