Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethylenediaminetetraacetic acid tetrasodium salt dihydrate | 10378-23-1 | MFCD00150026 | 100g
Catalog No. NC0962914
Encompass
Ethylenediaminetetraacetic acid tetrasodium salt dihydrate | Purity: 99.0-102.0% (titration) | MW:416.2 | 10378-23-1 | MFCD00150026 | 100g
Catalog No. NC0962914 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. ED4SS100G
Ethylenediaminetetraacetic acid tetrasodium salt dihydrate | Purity: 99.0-102.0% (titration) | MW:416.2 | 10378-23-1 | MFCD00150026 | 100g