Learn More
Medchemexpress LLC Edta calcium disodium | 62-33-9 | MFCD00148919 | 98.0% | 374.27 g/mol | C10H12CaN2Na2O8 | 100 G
SDP
Supplier:  Medchemexpress LLC HY107921100G
Encompass_Preferred
Edta calcium disodium is a chelating agent that binds divalent metal cations such as calcium, preventing metal-catalyzed reactions in biochemical and analytical workflows. Supplied as a white to off-white solid at 98% purity, it is suitable for preparing aqueous solutions and for use in analytical, industrial, and laboratory applications.
  • 98% purity suitable for analytical and laboratory use.
  • High water solubility for easy preparation of aqueous solutions.
  • Effective chelation of calcium and other divalent metal ions.
  • White to off-white solid with consistent appearance.
  • Stable when stored sealed and protected from moisture.
Catalog No. 50-002-42655