Learn More
Medchemexpress LLC Edta dipotassium dihydrate | 25102-12-9 | 99.17% | 50 G
SDP
Catalog No. 5000410098
Encompass_Preferred
EDTA dipotassium dihydrate is a metal ion chelator that forms stable and water-soluble chelates with a variety of metal ions. It is commonly used as a blood anticoagulant to prevent blood coagulation and maintain the morphology and function of cells in blood samples. In industrial applications, it can be used to remove adverse effects of metal ions and prevent oxidation and deterioration caused by metal ions.
  • Forms stable and water-soluble chelates with various metal ions.
  • Prevents blood coagulation, maintaining cell morphology and function in blood samples.
  • Removes adverse effects of metal ions in industrial settings.
  • Prevents oxidation and deterioration caused by metal ions.
  • Suitable for hematological testing.
Catalog No. 50-004-10098 Supplier Medchemexpress LLC Supplier No. HYD083650G
EDTA dipotassium dihydrate is a metal ion chelator that forms stable and water-soluble chelates with a variety of metal ions. It is commonly used as a blood anticoagulant to prevent blood coagulation and maintain the morphology and function of cells in blood samples. In industrial applications, it can be used to remove adverse effects of metal ions and prevent oxidation and deterioration caused by metal ions.
  • Forms stable and water-soluble chelates with various metal ions.
  • Prevents blood coagulation, maintaining cell morphology and function in blood samples.
  • Removes adverse effects of metal ions in industrial settings.
  • Prevents oxidation and deterioration caused by metal ions.
  • Suitable for hematological testing.