Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA dipotassium magnesium salt, For complexometry, ≥98%, Solstice

Catalog No. 6002433 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
6 x 100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-024-33 100 g
60-046-231 6 x 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-024-33 Supplier Solstice Supplier No. 33350100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for complexometry,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15708-48-2
Molecular Formula C10H12K2MgN2O8
Molecular Weight (g/mol) 390.714
MDL Number MFCD00054450
InChI Key MUEOBEUHFKBRJH-UHFFFAOYSA-J
Synonym dipotassium magnesium ethylenediaminetetraacetate, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-.kappa.o methyl glycinato-.kappa.n,.kappa.o 4--, dipotassium, oc-6-21, edta dipotassium magnesium salt monohydrate, ethylenediaminetetraacetic acid dipotassium magnesium salt, ethylenediamine-n,n,n',n'-tetraacetic acid dipotassium magnesium salt, edta, dipotassium magnesium salt, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-kappao methyl glycinato-kappan,kappao 4--, dipotassium, oc-6-21, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-kappao methyl glycinato-kappan,kappao 4--, potassium 1:2 , oc-6-21, magnesium 2+ dipotassium ion ion 4-edta, dipotassium n,n'-ethylenebis n-carboxymethyl glycinato 4--n,n',o,o',on,on' magnesate 2-
PubChem CID 85061
IUPAC Name magnesium;dipotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Mg+2].[K+].[K+]

Specifications

Physical Form Solid
Quantity 100 g
UN Number NONH for all modes of transport
Formula Weight 426.74g/mol
Percent Purity ≥98%
Grade Not Available
Chemical Name or Material Ethylenediaminetetraacetic acid dipotassium magnesium salt
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.