Learn More
Bioworld EDTA, Dipotassium Salt Dihydrate, 1 kg
Catalog No. 50255937
Encompass_Preferred
Used to eliminate inhibition of enzyme catalyzed reactions due to the trace presence of heavy metals.A metal chelating agent commonly used in tissue culture media and biological buffers.Specifications:Ultra Pure GradeChloride: <0.005%Iron: <0.001%Heavy metals (as Pb): <0.001%Appearance: LiquidHS Code : 29224985Melting Point: 255-280 °CPH Value: pH (5%, water, 250º°C): 4.0-5.0
Catalog No. 50-255-937 Supplier Bioworld Supplier No. 405000082
Used to eliminate inhibition of enzyme catalyzed reactions due to the trace presence of heavy metals.A metal chelating agent commonly used in tissue culture media and biological buffers.Specifications:Ultra Pure GradeChloride: <0.005%Iron: <0.001%Heavy metals (as Pb): <0.001%Appearance: LiquidHS Code : 29224985Melting Point: 255-280 °CPH Value: pH (5%, water, 250º°C): 4.0-5.0