Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Disodium, 0.0575 M Solution, Spectrum™ Chemical
SDP

Catalog No. 18610338 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Cubitainer
Poly Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-338 500 mL Poly Drum
18-610-336 1 L Poly Drum
18-610-337 4 L Poly Drum
18-610-334 20 L Cubitainer
18-610-335 200 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-610-338 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E115500MLPL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material EDTA Disodium
CAS 6381-92-6, 7732-18-5
CAS Max % 2.14, 97.86
Concentration 0.0575 M± 0.000 M
Grade Chemical Solution
Packaging Poly Drum
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.