Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA, Disodium, 0.100 Molar, Reagents

EDTA, Disodium, 0.100 Molar, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Supplier:  Reagents Holdings Llc C72011901A

Quantity 1 L

 View more versions of this product

Catalog No. 01-336-500


 Restricted Item
Add to Cart

Description

Description

Specifications

Chemical Identifiers

7732-18-5 , 6381-92-6
18.02
Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water

Specifications

Colorless
100°C
Edetate Disodium, Water Hardness Titrant, (Ethylenedinitrilo)tetraacetic Acid disodium salt
O
18.02
Poly Bottle
4 to 6
Non-sterile
EDTA, Disodium, 0.100 Molar
7732-18-5 , 6381-92-6
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Reagent
1.018
1 L
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.