Learn More
Medchemexpress LLC EDTA (disodium) | 139-33-3 | MFCD00070672 | 98.0% | 336.21 g/mol | C10H14N2Na2O8 | 10 G
SDP
Catalog No. 5000301072
Encompass_Preferred
EDTA disodium (ethylenediaminetetraacetic acid disodium salt) is a water-soluble chelating agent that sequesters divalent and trivalent metal ions. It is commonly used in biochemical, analytical, and formulation applications to control metal-catalyzed reactions, stabilize solutions, and improve product shelf life. The material is provided in small research pack sizes with a reported purity of 98.0%.
  • Water-soluble chelating agent for sequestering metal ions.
  • Helps prevent metal-catalyzed oxidation and degradation in formulations.
  • Suitable for buffer preparation and analytical methods.
  • Reported purity 98.0% for general laboratory use.
  • Available in small research pack sizes for convenience.
Catalog No. 50-003-01072 Supplier Medchemexpress LLC Supplier No. HYW01874610G
EDTA disodium (ethylenediaminetetraacetic acid disodium salt) is a water-soluble chelating agent that sequesters divalent and trivalent metal ions. It is commonly used in biochemical, analytical, and formulation applications to control metal-catalyzed reactions, stabilize solutions, and improve product shelf life. The material is provided in small research pack sizes with a reported purity of 98.0%.
  • Water-soluble chelating agent for sequestering metal ions.
  • Helps prevent metal-catalyzed oxidation and degradation in formulations.
  • Suitable for buffer preparation and analytical methods.
  • Reported purity 98.0% for general laboratory use.
  • Available in small research pack sizes for convenience.