Learn More
Medchemexpress LLC EDTA (disodium) | 139-33-3 | MFCD00150037 | 98.0% | 336.21 g/mol | C10H14N2Na2O8 | 25 G
SDP
Catalog No. 5000303867
Encompass_Preferred
Ethylenediaminetetraacetic acid disodium (EDTA disodium) is the disodium salt form of EDTA supplied as a white to off-white solid. It functions as a chelating agent to bind divalent and trivalent metal ions and is intended for laboratory research, analytical, and industrial applications. Storage guidance varies for solid and solution forms.
Catalog No. 50-003-03867 Supplier Medchemexpress LLC Supplier No. 25GHYW018746
Ethylenediaminetetraacetic acid disodium (EDTA disodium) is the disodium salt form of EDTA supplied as a white to off-white solid. It functions as a chelating agent to bind divalent and trivalent metal ions and is intended for laboratory research, analytical, and industrial applications. Storage guidance varies for solid and solution forms.