Learn More
Gold Biotechnology Inc EDTA Disodium dihydrate 1 kg
SDP
Catalog No. 5000562075
Encompass_Preferred
EDTA (Ethylenediaminetetraacetic acid) is a chelating agent a general chemical and a sequestrant In molecular biology applications it is used to minimize metal ion contamination and prevent enzymatic activity
Catalog No. 50-005-62075 Supplier Gold Biotechnology Inc Supplier No. E2101
EDTA (Ethylenediaminetetraacetic acid) is a chelating agent a general chemical and a sequestrant In molecular biology applications it is used to minimize metal ion contamination and prevent enzymatic activity