Learn More
Gold Biotechnology Inc EDTA Disodium dihydrate 500 g
SDP
Catalog No. 5000562156
Encompass_Preferred
EDTA (Ethylenediaminetetraacetic acid) is a chelating agent a general chemical and a sequestrant In molecular biology applications it is used to minimize metal ion contamination and prevent enzymatic activity
Catalog No. 50-005-62156 Supplier Gold Biotechnology Inc Supplier No. E210500
EDTA (Ethylenediaminetetraacetic acid) is a chelating agent a general chemical and a sequestrant In molecular biology applications it is used to minimize metal ion contamination and prevent enzymatic activity