Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Disodium Dihydrate, Purified ((Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate), Reagents

Catalog No. 01337259
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-259 500 g
01-337-261 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-259 Supplier Reagents Holdings Llc Supplier No. C2201100500A5
Add to Cart
Add to Cart

EDTA Disodium Dihydrate, Purified ((Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material EDTA Disodium Dihydrate, Purified ((Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate)
CAS 6381-92-6
Boiling Range >252°C
Melting Point 252°C
Color White
pH 4 to 6
Boiling Point >252°C
Melting Range 252°C
Packaging Poly Bottle
Molecular Formula C10H14N2O8Na2, 2H2O
Quantity 500 g
Specific Gravity 1
Synonym Edetate Disodium, Disodium EDTA, (Ethylenedinitrilo)tetraacetic Acid disodium salt dihydrate
Molecular Weight (g/mol) 372.2369
Grade Pure
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.