Learn More
AG Scientific Inc EDTA Disodium Salt, 250 G
Catalog No. 502237034
Encompass_Preferred
EDTA disodium salt, for molecular biology, is extensively used as a chelator of divalent cations such as Ca2+. EDTA is a chelating agent, a general chemical, and a sequestrant. In molecular biology applications, addition of EDTA minimizes the impact of metal ion contaminants. Also used to facilitate enzymatic reactions that could be inhibited by heavy metal traces and it also inhibits enzymes, such as metalloproteases.CAS Number: 6381-92-6Molecular Weight: 372.2 DaChemical Formula: C10H14N2O8Na2 2H2OSolubility: WaterStorage Temperature: Room TemperatureResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-7034 Supplier AG Scientific Inc Supplier No. E2483250GM
EDTA disodium salt, for molecular biology, is extensively used as a chelator of divalent cations such as Ca2+. EDTA is a chelating agent, a general chemical, and a sequestrant. In molecular biology applications, addition of EDTA minimizes the impact of metal ion contaminants. Also used to facilitate enzymatic reactions that could be inhibited by heavy metal traces and it also inhibits enzymes, such as metalloproteases.CAS Number: 6381-92-6Molecular Weight: 372.2 DaChemical Formula: C10H14N2O8Na2 2H2OSolubility: WaterStorage Temperature: Room TemperatureResearch or further manufacturing use only, not for food or drug use.