Learn More
Bioworld EDTA, Disodium Salt Dihydrate, 2.5 kg
Supplier:  Bioworld 405000123
Encompass_Preferred
EDTA, disodium salt, dihydrate A metal chelating agent commonly used in tissue culture media and biological buffers. Inactivates nucleases. Specifications:Also known as Ethylenediaminetetraacetate, disodium salt, dihydrateBiotechnology Grade (also available in Pretoemics Grade)Insolubles: <0.005%Heavy metals (as Pb): <0.005%Iron: <0.01%Nitrilotriacetic acid: <0.1%RNAse/DNAse activity: None detectedAppearance: PowderBoiling Point : 614.2 °C at 760 mmHg (Predicted)Density : 1.01 g/cm3 at 25 °CPH Value: pH (5%, water, 25ºC): 4.0-6.0RTECS Number: AH4410000
Catalog No. 50-255-940