Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences EDTA, Disodium Salt, Dihydrate, A.C.S. 100 GR
SDP

Catalog No. 50980463
Encompass_Preferred

(Ethylenediamine Tetraacetic acid)
CAS #6381-92-6 Na₂C₁₀H₁₄O8·₂H₂O F.W. 372.20
Assay: 99+%

Ethylenediamine Tetraacetic acid is used for the organic chelating for staining glutaraldehyde fixed tissues, pre-stained with uranyl acetate, followed by lead citrate. Bernard, W., J. Ultrastruct. Res., 27. 250 (1969)

Catalog No. 50-980-463 Supplier Electron Microscopy Sciences Supplier No. 15200
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Ethylenediamine Tetraacetic acid)
CAS #6381-92-6 Na₂C₁₀H₁₄O8·₂H₂O F.W. 372.20
Assay: 99+%

Ethylenediamine Tetraacetic acid is used for the organic chelating for staining glutaraldehyde fixed tissues, pre-stained with uranyl acetate, followed by lead citrate. Bernard, W., J. Ultrastruct. Res., 27. 250 (1969)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.