Learn More
Alkali Scientific EDTA Disodium Salt [Ethylenediaminetetra acetic acid, disodium salt dihydrate], 10 Kg
SDP
Catalog No. 5000661059
Encompass_Preferred
EDTA Disodium Salt (10 kg) is a highly concentrated chelating agent used for a variety of laboratory applications. Its ability to sequester metal ions makes it essential in preventing metal ion catalysis and ensuring accurate results in chemical reactions. This 10 kg bulk package is commonly used in molecular biology, biochemistry, and microbiology laboratories. EDTA effectively inhibits enzymes that require metal ions as cofactors, preventing undesired reactions during sample preparation, analysis, and preservation. This large volume is perfect for laboratories requiring frequent use of chelating agents in large-scale experiments or industrial applications, where controlling the concentration of metal ions is critical.
Catalog No. 50-006-61059 Supplier Alkali Scientific Supplier No. DTA002CP
EDTA Disodium Salt (10 kg) is a highly concentrated chelating agent used for a variety of laboratory applications. Its ability to sequester metal ions makes it essential in preventing metal ion catalysis and ensuring accurate results in chemical reactions. This 10 kg bulk package is commonly used in molecular biology, biochemistry, and microbiology laboratories. EDTA effectively inhibits enzymes that require metal ions as cofactors, preventing undesired reactions during sample preparation, analysis, and preservation. This large volume is perfect for laboratories requiring frequent use of chelating agents in large-scale experiments or industrial applications, where controlling the concentration of metal ions is critical.