Learn More
Alkali Scientific EDTA Disodium Salt [Ethylenediaminetetra acetic acid, disodium salt dihydrate], 2.5 Kg
SDP
Catalog No. 5000661061
Encompass_Preferred
EDTA Disodium Salt (2.5 kg) is a high-concentration chelating agent used for laboratory experiments that require the removal or sequestration of metal ions. This 2.5 kg package is ideal for large-scale applications in biochemistry, molecular biology, and other scientific fields where metal ion control is essential. EDTA works by binding to metal ions such as calcium, magnesium, and iron, preventing them from catalyzing unwanted chemical reactions or interfering with enzymatic activity. It is widely used in DNA and RNA extraction, enzyme assays, and metal ion quantification. Laboratories that conduct frequent experiments requiring consistent metal ion control benefit from this bulk packaging.
Catalog No. 50-006-61061 Supplier Alkali Scientific Supplier No. DTA004CP
EDTA Disodium Salt (2.5 kg) is a high-concentration chelating agent used for laboratory experiments that require the removal or sequestration of metal ions. This 2.5 kg package is ideal for large-scale applications in biochemistry, molecular biology, and other scientific fields where metal ion control is essential. EDTA works by binding to metal ions such as calcium, magnesium, and iron, preventing them from catalyzing unwanted chemical reactions or interfering with enzymatic activity. It is widely used in DNA and RNA extraction, enzyme assays, and metal ion quantification. Laboratories that conduct frequent experiments requiring consistent metal ion control benefit from this bulk packaging.