Learn More
Alkali Scientific EDTA Disodium Salt [Ethylenediaminetetra acetic acid, disodium salt dihydrate], 250 G
SDP
Catalog No. 5000661060
Encompass_Preferred
EDTA Disodium Salt (250 g) is a versatile chelating agent used in various laboratory applications to bind metal ions and prevent metal-catalyzed reactions. This reagent is commonly used in molecular biology, biochemistry, and analytical chemistry for tasks like DNA extraction, protein purification, and enzyme inhibition studies. EDTA functions by sequestering divalent and trivalent metal ions, preventing them from interfering with chemical reactions or biological processes. Its ability to stabilize samples by controlling metal ion concentration makes it an essential component in laboratory research, clinical diagnostics, and various analytical techniques that require metal ion control.
Catalog No. 50-006-61060 Supplier Alkali Scientific Supplier No. DTA003CP
EDTA Disodium Salt (250 g) is a versatile chelating agent used in various laboratory applications to bind metal ions and prevent metal-catalyzed reactions. This reagent is commonly used in molecular biology, biochemistry, and analytical chemistry for tasks like DNA extraction, protein purification, and enzyme inhibition studies. EDTA functions by sequestering divalent and trivalent metal ions, preventing them from interfering with chemical reactions or biological processes. Its ability to stabilize samples by controlling metal ion concentration makes it an essential component in laboratory research, clinical diagnostics, and various analytical techniques that require metal ion control.