Learn More
Alkali Scientific EDTA Disodium Salt [Ethylenediaminetetra acetic acid, disodium salt dihydrate], 500 G
SDP
Catalog No. 5000660226
Encompass_Preferred
EDTA Disodium Salt (500 g) is a reliable chelating agent that is essential for controlling metal ions in laboratory settings. This reagent is commonly used in molecular biology and biochemistry experiments to prevent metal-catalyzed reactions that could compromise experimental results. EDTA works by forming stable complexes with metal ions, preventing them from interacting with other compounds in the sample. It is particularly useful in DNA extractions, enzyme inhibition studies, and protein purification protocols. The 500 g size is perfect for small-to-medium-scale laboratory use, making it a cost-effective solution for frequent metal ion control needs in research applications.
Catalog No. 50-006-60226 Supplier Alkali Scientific Supplier No. DTA005CP
EDTA Disodium Salt (500 g) is a reliable chelating agent that is essential for controlling metal ions in laboratory settings. This reagent is commonly used in molecular biology and biochemistry experiments to prevent metal-catalyzed reactions that could compromise experimental results. EDTA works by forming stable complexes with metal ions, preventing them from interacting with other compounds in the sample. It is particularly useful in DNA extractions, enzyme inhibition studies, and protein purification protocols. The 500 g size is perfect for small-to-medium-scale laboratory use, making it a cost-effective solution for frequent metal ion control needs in research applications.