Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA disodium salt solution, Volumetric, Reag. Ph. Eur., 0.1 M EDTA-Na2, for complexometry, Solstice

Catalog No. p-7072116 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-025-39 500 mL
60-025-37 1 L
60-025-38 2.5 L
60-025-36 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-025-39 Supplier Solstice Supplier No. 34550500ML
Add to Cart
Add to Cart

Volumetric, Reag. Ph. Eur., 0.1 M EDTA-Na2, for complexometry

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethylenediaminetetraacetic acid disodium salt solution
CAS 139-33-3
Molecular Formula C10H14N2Na2O8
Linear Formula [-CH2N(CH2CO2Na)CH2CO2H]2
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
UN Number NONH for all modes of transport
Beilstein 3822669
Synonym ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
IUPAC Name disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
Molecular Weight (g/mol) 336.21
PubChem CID 57339238
ChEBI CHEBI:64734
Formula Weight 336.21g/mol
Quantity 500 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.