Learn More
Research Products International Corp EDTA Ferric Sodium Salt [Ethylenediaminetetraacetic acid ferric sodium salt], 250 Grams
SDP
Supplier:  Research Products International Corp E57040250.0
Encompass_Preferred
Purity: >99%. Soluble in: Water. Store at: Room Temperature. CAS No. 15708-41-5
Catalog No. 50-213-303