Learn More
Alkali Scientific EDTA Ferric Sodium Salt [Ethylenediaminetetraacetic acid ferric sodium salt], 1 Kg
SDP
Catalog No. 5000660991
Encompass_Preferred
EDTA Ferric Sodium Salt (1 kg) is a high-purity chelating agent commonly used in research to form stable iron complexes. This reagent plays a crucial role in experiments requiring precise control over metal ion concentrations, as it binds to iron and other metal ions, preventing them from catalyzing unwanted reactions. EDTA Ferric Sodium Salt is essential in biological research, including protein analysis and enzymatic assays, where metal ion interference must be minimized. It is also used in metal sequestration in environmental studies and water purification. The 1 kg packaging is ideal for laboratories conducting high-throughput or ongoing research requiring significant quantities of this compound.
Catalog No. 50-006-60991 Supplier Alkali Scientific Supplier No. DTA013CP
EDTA Ferric Sodium Salt (1 kg) is a high-purity chelating agent commonly used in research to form stable iron complexes. This reagent plays a crucial role in experiments requiring precise control over metal ion concentrations, as it binds to iron and other metal ions, preventing them from catalyzing unwanted reactions. EDTA Ferric Sodium Salt is essential in biological research, including protein analysis and enzymatic assays, where metal ion interference must be minimized. It is also used in metal sequestration in environmental studies and water purification. The 1 kg packaging is ideal for laboratories conducting high-throughput or ongoing research requiring significant quantities of this compound.