Learn More
Alkali Scientific EDTA Ferric Sodium Salt [Ethylenediaminetetraacetic acid ferric sodium salt], 100 G
SDP
Catalog No. 5000659844
Encompass_Preferred
EDTA Ferric Sodium Salt is a chelating agent used for iron complexation in laboratory applications. Available in a 100g quantity, this compound is frequently used in research where metal ions need to be tightly controlled or removed from solutions. The EDTA structure binds to metal ions like iron, preventing their interaction in enzymatic reactions and other biochemical processes. This reagent is commonly used in metal ion sequestration experiments, biological assays, and iron-related research studies. It is also valuable in protein purification processes where iron contamination can interfere with results. EDTA Ferric Sodium Salt provides consistent performance for a variety of analytical and chemical processes.
Catalog No. 50-006-59844 Supplier Alkali Scientific Supplier No. DTA012CP
EDTA Ferric Sodium Salt is a chelating agent used for iron complexation in laboratory applications. Available in a 100g quantity, this compound is frequently used in research where metal ions need to be tightly controlled or removed from solutions. The EDTA structure binds to metal ions like iron, preventing their interaction in enzymatic reactions and other biochemical processes. This reagent is commonly used in metal ion sequestration experiments, biological assays, and iron-related research studies. It is also valuable in protein purification processes where iron contamination can interfere with results. EDTA Ferric Sodium Salt provides consistent performance for a variety of analytical and chemical processes.