Learn More
Bioworld EDTA, Ferric Sodium Salt, 1 kg
Catalog No. 50255943
Encompass_Preferred
An eliminator of enzyme catalyzed reaction inhibition. Plant culture tested. Supports or facilitates plant growth and/or shoot proliferation in two or more plant tissue cultures.Appearance: Yellow to Brown Crystalline PowderBoiling Point : 614.2°CDensity : 7.87 g/cm3 at 25 °CIUPAC_Name: iron(3+) ion sodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatome)Melting Point: 80 °CRTECS Number: AH4900000
Catalog No. 50-255-943 Supplier Bioworld Supplier No. 405000163
An eliminator of enzyme catalyzed reaction inhibition. Plant culture tested. Supports or facilitates plant growth and/or shoot proliferation in two or more plant tissue cultures.Appearance: Yellow to Brown Crystalline PowderBoiling Point : 614.2°CDensity : 7.87 g/cm3 at 25 °CIUPAC_Name: iron(3+) ion sodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatome)Melting Point: 80 °CRTECS Number: AH4900000