Learn More
Braintree Scientific Prepared Microtubes with Push Cap,1.3ml, EDTA, 100/bx
SDP
Catalog No. NC0299959
Encompass
Prepared Micro Tubes with Attached Push Cap Closure. Skirted, conical base and label. Preparations: Serum clotting activator, plasma lithium heparin, hematology potassium EDTA and coagulation citrate **Sold for research purposes only and not for human use**
Catalog No. NC0299959 Supplier Braintree Scientific Supplier No. PMT1504PK
Prepared Micro Tubes with Attached Push Cap Closure. Skirted, conical base and label. Preparations: Serum clotting activator, plasma lithium heparin, hematology potassium EDTA and coagulation citrate **Sold for research purposes only and not for human use**