Learn More
Hach Company EDTA Reagent Powder Pillows, pk/100
Supplier:  Hach Company 700599
Encompass
For determination of cobalt and nickel by the PAN method (Hach methods 8078 and 8150). Also used in Phenols test kits. Pack of 100 powder pillows.
Catalog No. 50-199-4823