Learn More
Braintree Scientific Minivette POCT, 100ul, EDTA, red, 200/pk
SDP
Catalog No. NC1569776
Encompass
Minivette POCT - Easy sample recovery due to optimal capillary features. Precise dispensing of small whole blood volumes. Prevents spillage during transfer to test card. Available in 20ul and 50ul volumes. Preparations: Neutral, heparin and EDTA **Sold for research purposes only and not for human use**
Catalog No. NC1569776 Supplier Braintree Scientific Supplier No. MP1002113PK
Minivette POCT - Easy sample recovery due to optimal capillary features. Precise dispensing of small whole blood volumes. Prevents spillage during transfer to test card. Available in 20ul and 50ul volumes. Preparations: Neutral, heparin and EDTA **Sold for research purposes only and not for human use**