Learn More
EDTA (Sodium Edetate), 0.100 M EP Volumetric Solution 3005900, Reagents
EDTA (Sodium Edetate), 0.100 M EP Volumetric Solution 3005900, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$83.40
Specifications
| Chemical Name or Material | EDTA (Sodium Edetate), 0.100 M EP Volumetric Solution 3005900 |
|---|---|
| Color | Colorless |
| CAS | 7732-18-5 , 6381-92-6 |
| Boiling Point | 100°C |
| Molecular Formula | C10H14N2O8Na2/H2O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-335-390
|
Reagents Holdings Llc
CE0030001A |
1 L |
Each of 1 for $83.40
|
|
|||||
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.