Learn More
Hach Company Hach EDTA Solution 0.005 N, 2 L for EZ4043 Total Hardness Analyzer
Catalog No. NC2227580 Shop All Hach Company Products
Encompass
Hach EDTA solution APPC4043-04 is a ready-to-use titration reagent formulated at 0.005 N for automated total hardness measurement. Supplied in a 2 L container, it provides the EDTA titrant used for calcium and magnesium determination in online total hardness analyzers.
  • Ready-to-use EDTA titration reagent.
  • Concentration: 0.005 N.
  • Volume: 2 L.
  • Intended for total hardness titration in automated analyzers.
This item is not returnable. View return policy
Catalog No. NC2227580 Supplier Hach Company Supplier No. APPC404304
Hach EDTA solution APPC4043-04 is a ready-to-use titration reagent formulated at 0.005 N for automated total hardness measurement. Supplied in a 2 L container, it provides the EDTA titrant used for calcium and magnesium determination in online total hardness analyzers.
  • Ready-to-use EDTA titration reagent.
  • Concentration: 0.005 N.
  • Volume: 2 L.
  • Intended for total hardness titration in automated analyzers.