Learn More
IBI SCIENTIFIC 3P (EDTA) SOLUTION, 100 mL
Catalog No. NC1830198
Encompass_Preferred
IBI 0.5MEDTA solution (pH 8.0)is ideal for biochemistry and molecular biology applications requiring a chelator of divalent metal ions. Ethylenediamine tetraacetic acid solution can be used to elucidate the role of antigen retrieval in immunostaining of ethanol fixed smears.
Catalog No. NC1830198 Supplier IBI SCIENTIFIC 3P Supplier No. IB70184
IBI 0.5MEDTA solution (pH 8.0)is ideal for biochemistry and molecular biology applications requiring a chelator of divalent metal ions. Ethylenediamine tetraacetic acid solution can be used to elucidate the role of antigen retrieval in immunostaining of ethanol fixed smears.