Learn More
Hach Company EDTA Standard Solution, 0.0200N, 4L
Supplier:  Hach Company 20556
Encompass
For hardness determination by buret titration. Hach methods 8222 (calcium) and 8226 (total). 0.0200N (0.0100 M) (N/50) 4 liter bottle
Catalog No. 50-199-4208