Learn More
Malvern Panalytical EDTA Test Kit ITC200, at least 10 sings runs
Catalog No. NC0626823
Encompass
EDTA Test Kit Manual ITC200. This kit allows users to test instrument performance using EDTA. Expect at least 10 single runs of material from this kit. This kit will ship with at least 90 days of use before material expiry.This item has a minimum qty of 2 per supplier requirements
Catalog No. NC0626823 Supplier Malvern Panalytical Supplier No. 10048368
EDTA Test Kit Manual ITC200. This kit allows users to test instrument performance using EDTA. Expect at least 10 single runs of material from this kit. This kit will ship with at least 90 days of use before material expiry.This item has a minimum qty of 2 per supplier requirements