Learn More
Bioworld EDTA, TetraSodium Salt Dihydrate, 2.5 kg
Supplier:  Bioworld 405000243
Encompass_Preferred
A metal chelating agent commonly used in tissue culture media and biological buffers. Technical Information:Also known as Ethylenediaminetetraacetate, tetrasodium salt, dihydrateUltra Pure GradeHeavy metals (as Pb): <0.001%Iron: <0.005%Nitrilotriacetic acid: <0.1%Appearance: Powder or crystalline or crystalline powderPH Value: •pH (5%, water, 25ºC): 10.7-11.7
Catalog No. 50-255-948