Learn More
HIMEDIA EDTA Tetrasodium Salt Dihydrate, Hi-LR, GRM6352, Laboratory Chemicals, Chelating Agent, Laboratory Reagent Grade, CAS 10378-23-1, Complexometric Reagent, Powder, 100 g
Catalog No. NC4013653
Encompass
EDTA tetrasodium salt dihydrate, Hi-LR is a laboratory reagent grade chelating agent (C10H12N2Na4O8.2H2O, CAS 10378-23-1, molecular weight 416.20) supplied as a readily water-soluble white crystalline powder. Its strong, well-defined affinity for metal ions makes it a workhorse for sequestering calcium, magnesium and heavy metals in buffers, complexometric titrations, cleaning solutions and molecular biology reagents. Store below 30 C, shelf life four years. Packaged as 100 g.
Catalog No. NC4013653 Supplier HIMEDIA Supplier No. GRM6352100G
EDTA tetrasodium salt dihydrate, Hi-LR is a laboratory reagent grade chelating agent (C10H12N2Na4O8.2H2O, CAS 10378-23-1, molecular weight 416.20) supplied as a readily water-soluble white crystalline powder. Its strong, well-defined affinity for metal ions makes it a workhorse for sequestering calcium, magnesium and heavy metals in buffers, complexometric titrations, cleaning solutions and molecular biology reagents. Store below 30 C, shelf life four years. Packaged as 100 g.